Merck
  • Inositol triphosphate-triggered calcium release from the endoplasmic reticulum induces lysosome biogenesis via TFEB/TFE3.
Inositol triphosphate-triggered calcium release from the endoplasmic reticulum induces lysosome biogenesis via TFEB/TFE3.
The Journal of biological chemistry (2022-02-20)
Mouhannad Malek, Anna M Wawrzyniak, Michael Ebner, Dmytro Puchkov, Volker Haucke
ABSTRACT
Lysosomes serve as dynamic regulators of cell and organismal physiology by integrating the degradation of macromolecules with receptor and nutrient signaling. Previous studies have established that activation of the transcription factor EB (TFEB) and transcription factor E3 (TFE3) induces the expression of lysosomal genes and proteins in signaling-inactive starved cells, that is, under conditions when activity of the master regulator of nutrient-sensing signaling mechanistic target of rapamycin complex 1 is repressed. How lysosome biogenesis is triggered in signaling-active cells is incompletely understood. Here, we identify a role for calcium release from the lumen of the endoplasmic reticulum in the control of lysosome biogenesis that is independent of mechanistic target of rapamycin complex 1. We show using functional imaging that calcium efflux from endoplasmic reticulum stores induced by inositol triphosphate accumulation upon depletion of inositol polyphosphate-5-phosphatase A, an inositol 5-phosphatase downregulated in cancer and defective in spinocerebellar ataxia, or receptor-mediated phospholipase C activation leads to the induction of lysosome biogenesis. This mechanism involves calcineurin and the nuclear translocation and elevated transcriptional activity of TFEB/TFE3. Our findings reveal a crucial function for inositol polyphosphate-5-phosphatase A-mediated triphosphate hydrolysis in the control of lysosome biogenesis via TFEB/TFE3, thereby contributing to our understanding how cells are able to maintain their lysosome content under conditions of active receptor and nutrient signaling.
Sigma-Aldrich
Gö 6976, Gö 6976, CAS 136194-77-9, is a cell-permeable, reversible, and ATP-competitive inhibitor of PKC (IC₅₀ = 7.9 nM for rat brain). Exhibits selectively for PKCα (IC₅₀ = 2.3 nM) and βI (IC₅₀ = 6.2 nM).
Sigma-Aldrich
Phospholipase C Activator, m-3M3FBS, The Phospholipase C Activator, m-3M3FBS, also referenced under CAS 200933-14-8, controls the biological activity of Phospholipase C. This small molecule/inhibitor is primarily used for Activators/Inducers applications.