Merck
  • DNA damage in embryonic neural stem cell determines FTLDs' fate via early-stage neuronal necrosis.
DNA damage in embryonic neural stem cell determines FTLDs' fate via early-stage neuronal necrosis.
Life science alliance (2021-06-17)
Hidenori Homma, Hikari Tanaka, Meihua Jin, Xiaocen Jin, Yong Huang, Yuki Yoshioka, Christian Jf Bertens, Kohei Tsumaki, Kanoh Kondo, Hiroki Shiwaku, Kazuhiko Tagawa, Hiroyasu Akatsu, Naoki Atsuta, Masahisa Katsuno, Katsutoshi Furukawa, Aiko Ishiki, Masaaki Waragai, Gaku Ohtomo, Atsushi Iwata, Takanori Yokota, Haruhisa Inoue, Hiroyuki Arai, Gen Sobue, Masaki Sone, Kyota Fujita, Hitoshi Okazawa
ABSTRACT
The early-stage pathologies of frontotemporal lobal degeneration (FTLD) remain largely unknown. In VCPT262A-KI mice carrying VCP gene mutation linked to FTLD, insufficient DNA damage repair in neural stem/progenitor cells (NSCs) activated DNA-PK and CDK1 that disabled MCM3 essential for the G1/S cell cycle transition. Abnormal neural exit produced neurons carrying over unrepaired DNA damage and induced early-stage transcriptional repression-induced atypical cell death (TRIAD) necrosis accompanied by the specific markers pSer46-MARCKS and YAP. In utero gene therapy expressing normal VCP or non-phosphorylated mutant MCM3 rescued DNA damage, neuronal necrosis, cognitive function, and TDP43 aggregation in adult neurons of VCPT262A-KI mice, whereas similar therapy in adulthood was less effective. The similar early-stage neuronal necrosis was detected in PGRNR504X-KI, CHMP2BQ165X-KI, and TDPN267S-KI mice, and blocked by embryonic treatment with AAV-non-phospho-MCM3. Moreover, YAP-dependent necrosis occurred in neurons of human FTLD patients, and consistently pSer46-MARCKS was increased in cerebrospinal fluid (CSF) and serum of these patients. Collectively, developmental stress followed by early-stage neuronal necrosis is a potential target for therapeutics and one of the earliest general biomarkers for FTLD.
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, purified from hybridoma cell culture
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Tbr2 Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-DAP-Kinase (pSer308) antibody, Mouse monoclonal, clone DKPS308, purified from hybridoma cell culture
Sigma-Aldrich
CDK2/Cyclin A2, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Pan Cadherin antibody produced in mouse, clone CH-19, ascites fluid
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3 antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit