Merck
  • Filamin A is required for somatostatin receptor type 5 expression and pasireotide-mediated signaling in pituitary corticotroph tumor cells.
Filamin A is required for somatostatin receptor type 5 expression and pasireotide-mediated signaling in pituitary corticotroph tumor cells.
Molecular and cellular endocrinology (2021-01-12)
Donatella Treppiedi, Genesio Di Muro, Federica Mangili, Rosa Catalano, Elena Giardino, Anna Maria Barbieri, Marco Locatelli, Maura Arosio, Anna Spada, Erika Peverelli, Giovanna Mantovani
ABSTRACT
Somatostatin receptor type 5 (SST5) represents the main pharmacological target in the treatment of adrenocorticotroph hormone (ACTH)-secreting tumors. However, molecular predictors of responsiveness to pasireotide require further investigation. The cytoskeleton protein filamin A (FLNA) modulates the responsiveness to somatostatin analogs (SSA) treatment in other types of pituitary tumors by regulating somatostatin receptor type 2 (SST2)/dopamine receptor type 2 (DRD2) expression and activity. Here, we aimed to test the involvement of FLNA in the modulation of SST5 response to SSA in human and murine tumor corticotrophs. Western blot analysis of human corticotropinomas showed that FLNA and SST5 correlate. Both in human primary cultures and AtT-20 cells, FLNA genetic silencing caused a decrease of receptor expression level. Moreover, pasireotide-mediated SST5 downregulation observed in AtT-20 control cells was no further detected in FLNA silenced cells. In AtT-20 cells, in situ PLA experiments revealed an increased number of SST5-FLNA complexes following pasireotide incubation. Finally, FLNA knock down abolished pasireotide-induced SST5 actions on hormone secretion, cell proliferation and apoptosis. In conclusion, FLNA is implicated in SST5 expression modulation and signaling.
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting human FLNA