Merck
  • Glutamine Antagonist JHU-083 Normalizes Aberrant Hippocampal Glutaminase Activity and Improves Cognition in APOE4 Mice.
Glutamine Antagonist JHU-083 Normalizes Aberrant Hippocampal Glutaminase Activity and Improves Cognition in APOE4 Mice.
Journal of Alzheimer's disease : JAD (2020-07-18)
Kristen R Hollinger, Xiaolei Zhu, Elizabeth S Khoury, Ajit G Thomas, Kevin Liaw, Carolyn Tallon, Ying Wu, Eva Prchalova, Atsushi Kamiya, Camilo Rojas, Sujatha Kannan, Barbara S Slusher
ABSTRACT
Given the emergent aging population, the identification of effective treatments for Alzheimer's disease (AD) is critical. We investigated the therapeutic efficacy of JHU-083, a brain-penetrable glutamine antagonist, in treating AD using the humanized APOE4 knock-in mouse model. Cell culture studies were performed using BV2 cells and primary microglia isolated from hippocampi of adult APOE4 knock-in mice to evaluate the effect of JHU-083 treatment on LPS-induced glutaminase (GLS) activity and inflammatory markers. Six-month-old APOE4 knock-in mice were administered JHU-083 or vehicle via oral gavage 3x/week for 4-5 months and cognitive performance was assessed using the Barnes maze. Target engagement in the brain was confirmed using a radiolabeled GLS enzymatic activity assay, and electrophysiology, gastrointestinal histology, blood chemistry, and CBC analyses were conducted to evaluate the tolerability of JHU-083. JHU-083 inhibited the LPS-mediated increases in GLS activity, nitic oxide release, and pro-inflammatory cytokine production in cultured BV2 cells and primary microglia isolated from APOE4 knock-in AD mice. Chronic treatment with JHU-083 in APOE4 mice improved hippocampal-dependent Barnes maze performance. Consistent with the cell culture findings,postmortem analyses of APOE4 mice showed increased GLS activity in hippocampal CD11b+ enriched cells versus age-matched controls, which was completely normalized by JHU-083 treatment. JHU-083 was well-tolerated, showing no weight loss effect or overt behavioral changes. Peripheral nerve function, gastrointestinal histopathology, and CBC/clinical chemistry parameters were all unaffected by chronic JHU-083 treatment. These results suggest that the attenuation of upregulated hippocampal glutaminase by JHU-083 represents a new therapeutic strategy for AD.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O127:B8, purified by ion-exchange chromatography, TLR ligand tested
Pricing and availability is not currently available.