Merck
  • Biomimetic nanofibrous gelatin/apatite composite scaffolds for bone tissue engineering.
Biomimetic nanofibrous gelatin/apatite composite scaffolds for bone tissue engineering.
Biomaterials (2009-01-21)
Xiaohua Liu, Laura A Smith, Jiang Hu, Peter X Ma
ABSTRACT
Mimicking certain features (e.g. nanoscale topography and biological cues) of natural extracellular matrix (ECM) is advantageous for the successful regeneration of damaged tissue. In this study, nanofibrous gelatin/apatite (NF-gelatin/apatite) composite scaffolds have been fabricated to mimic both the physical architecture and chemical composition of natural bone ECM. A thermally induced phase separation (TIPS) technique was developed to prepare nanofibrous gelatin (NF-gelatin) matrix. The NF-gelatin matrix mimicked natural collagen fibers and had an average fiber diameter of about 150nm. By integrating the TIPS method with porogen leaching, three-dimensional NF-gelatin scaffolds with well-defined macropores were fabricated. In comparison to Gelfoam (a commercial gelatin foam) with similar pore size and porosity, the NF-gelatin scaffolds exhibited a much higher surface area and mechanical strength. The surface area and compressive modulus of NF-gelatin scaffolds were more than 700 times and 10 times higher than that of Gelfoam, respectively. The NF-gelatin scaffolds also showed excellent biocompatibility and mechanical stability. To further enhance pre-osteoblast cell differentiation as well as improving mechanical strength, bone-like apatite particles (<2microm) were incorporated onto the surface of NF-gelatin scaffolds via a simulated body fluid (SBF) incubation process. The NF-gelatin/apatite scaffolds 5 days after SBF treatment showed significantly higher mechanical strength than NF-gelatin scaffolds 5 days after SBF treatment. Furthermore, the incorporated apatite in the NF-gelatin/apatite composite scaffold enhanced the osteogenic differentiation. The expression of BSP and OCN in the osteoblast-(NF-gelatin/apatite composite) constructs was about 5 times and 2 times higher than in the osteoblast-(NF-gelatin) constructs 4 weeks after cell culture. The biomimetic NF-gelatin/apatite scaffolds are, therefore, excellent for bone tissue engineering.
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra