Merck
  • Glycogenolysis Is Crucial for Astrocytic Glycogen Accumulation and Brain Damage after Reperfusion in Ischemic Stroke.
Glycogenolysis Is Crucial for Astrocytic Glycogen Accumulation and Brain Damage after Reperfusion in Ischemic Stroke.
iScience (2020-05-24)
Yanhui Cai, Haiyun Guo, Ze Fan, Xinlei Zhang, Di Wu, Wenhong Tang, Tingting Gu, Shiquan Wang, Anqi Yin, Liang Tao, Xunming Ji, Hailong Dong, Yan Li, Lize Xiong
ABSTRACT
Astrocytic glycogen is an important energy reserve in the brain and is believed to supply fuel during energy crisis. However, the pattern of glycogen metabolism in ischemic stroke and its potential therapeutic impact on neurological outcomes are still unknown. Here, we found extensive brain glycogen accumulation after reperfusion in ischemic stroke patients and primates. Glycogenolytic dysfunction in astrocytes is responsible for glycogen accumulation, caused by inactivation of the protein kinase A (PKA)-glycogen phosphorylase kinase (PhK)-glycogen phosphorylase (GP) cascade accompanied by the activation of glycogen synthase kinase-3β (GSK3β). Genetic or pharmacological augmentation of astrocytic GP could promote astrocyte and neuron survival and improve neurological behaviors. In addition, we found that insulin exerted a neuroprotective effect, at least in part by rescuing the PKA-PhK-GP cascade to maintain homeostasis of glycogen metabolism during reperfusion. Together, our findings suggest a promising intervention for undesirable outcomes in ischemic stroke.
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 1,000-5,000
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Arabitol, ≥99% (GC)
Sigma-Aldrich
Cell Counting Kit - 8, for quantitation of viable cell number in proliferation and cytotoxicity assays
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Cresyl Violet acetate, certified by the BSC