Merck
  • A Cross-Reactive Small Protein Binding Domain Provides a Model to Study Off-Tumor CAR-T Cell Toxicity.
A Cross-Reactive Small Protein Binding Domain Provides a Model to Study Off-Tumor CAR-T Cell Toxicity.
Molecular therapy oncolytics (2020-05-06)
Joanne A Hammill, Jacek M Kwiecien, Anna Dvorkin-Gheva, Vivian W C Lau, Christopher Baker, Ying Wu, Ksenia Bezverbnaya, Craig Aarts, Christopher W Heslen, Galina F Denisova, Heather Derocher, Katy Milne, Brad H Nelson, Jonathan L Bramson
ABSTRACT
Tumor-targeted chimeric antigen receptor (CAR)-engineered T lymphocytes (CAR-T cells) have demonstrated striking clinical success, but their use has been associated with a constellation of toxicities. A better understanding of the pathogenesis of these toxicities is required to improve the safety profile of CAR-T cells. Herein, we describe a xenograft model of off-tumor CAR-T cell-associated toxicity. Human CAR-T cells targeted against HER2 using a small-protein binding domain induced acute, dose-dependent toxicities in mice. The inclusion of a CD28 or 4-1BB co-stimulatory domain in the CAR was required to produce toxicity; however, co-stimulation through CD28 was most toxic on a per-cell basis. CAR-T cell activation in the lungs and heart was associated with a systemic cytokine storm. The severity of observed toxicities was dependent upon the peripheral blood mononuclear cell (PBMC) donor used as a T cell source and paralleled the CD4+-to-CD8+ T cell ratio in the adoptive transfer product. CD4+ CAR-T cells were determined to be the primary contributors to CAR-T cell-associated toxicity. However, donor-specific differences persisted after infusion of a purified CD4+ CAR-T cell product, indicating a role for additional variables. This work highlights the contributions of CAR-T cell-intrinsic variables to the pathogenesis of off-tumor toxicity.
Sigma-Aldrich
Monoclonal Anti-Cytokeratin, pan antibody produced in mouse, ascites fluid, clone PCK-26