Merck
  • Comparison of real wastewater oxidation with Fenton/Fenton-like and persulfate activated by NaOH and Fe(II).
Comparison of real wastewater oxidation with Fenton/Fenton-like and persulfate activated by NaOH and Fe(II).
Journal of environmental management (2020-02-18)
Sergio Rodríguez, David Lorenzo, Aurora Santos, Arturo Romero
ABSTRACT
Treatment of polluted wastewaters from industrial activities has become a source of major concern for the environment. In this work, real wastewater from a physico-chemical (WWFQ) treatment was tested through different oxidation technologies: Fenton and Fenton-like reagent and persulfate activated by NaOH and Fe(II). Oxidation reactions with Fenton's reagent were carried out in a 0.25 L batch reactor at 25 °C by adding either Fe(II) or Fe(III) and H2O2 to an aqueous solution of wastewater, whose pH was previously adjusted to 2 or 3. Iron concentration ranging from 25 to 100 mg/L and peroxide concentration from 2500 to 10000 mg/L were used. The total organic carbon slightly decreased when WWFQ was treated. Moreover, better results were obtained when Fe(II) was used than Fe(III). Both iron concentration and oxidant dosage had a positive influence on the chemical oxygen demand (COD) removal, until an asymptotic value of 30% was obtained. Oxidation of pollutants contained in WWFQ was studied with persulfate (18.4-294 mM) activated with NaOH and Fe(II) (36.8-588 mM). Again, a positive influence of both persulfate and NaOH was observed, although a similar asymptotic COD value was observed. This parallelism between both technologies confirms recalcitrant compounds were obtained.
Sigma-Aldrich
2,4,7,9-Tetramethyl-5-decyne-4,7-diol, mixture of (±) and meso, 98%