Merck
  • Ylide-Functionalized Phosphine (YPhos)-Palladium Catalysts: Selective Monoarylation of Alkyl Ketones with Aryl Chlorides.
Ylide-Functionalized Phosphine (YPhos)-Palladium Catalysts: Selective Monoarylation of Alkyl Ketones with Aryl Chlorides.
Organic letters (2019-08-31)
Xiao-Qiang Hu, Dominik Lichte, Ilja Rodstein, Philip Weber, Ann-Katrin Seitz, Thorsten Scherpf, Viktoria H Gessner, Lukas J Gooßen
ABSTRACT
Ylide-functionalized phosphine (YPhos) ligands allow the palladium-catalyzed α-arylation of alkyl ketones with aryl chlorides with record setting activity. Using a cyclohexyl-substituted YPhos ligand, a wide range of challenging ketone substrates was efficiently and selectively monoarylated under mild conditions. A newly designed YPhos ligand bearing tert-butyl groups on the coordinating phosphorus atom is already active at room temperature. The synthetic potential was demonstrated by gram-scale reactions and the succinct synthesis of ε-caprolactone derivatives.
Sigma-Aldrich
joYPhos, Umicore
Sigma-Aldrich
keYPhos
Sigma-Aldrich
trYPhos, Umicore