Merck
  • Comparative analysis of TTF-1 binding DNA regions in small-cell lung cancer and non-small-cell lung cancer.
Comparative analysis of TTF-1 binding DNA regions in small-cell lung cancer and non-small-cell lung cancer.
Molecular oncology (2019-11-30)
Satoshi Hokari, Yusuke Tamura, Atsushi Kaneda, Akihiro Katsura, Masato Morikawa, Fumihiko Murai, Shogo Ehata, Shuichi Tsutsumi, Yuichi Ishikawa, Hiroyuki Aburatani, Toshiaki Kikuchi, Kohei Miyazono, Daizo Koinuma
ABSTRACT
Thyroid transcription factor-1 (TTF-1, encoded by the NKX2-1 gene) is highly expressed in small-cell lung carcinoma (SCLC) and lung adenocarcinoma (LADC), but how its functional roles differ between SCLC and LADC remains to be elucidated. Here, we compared the genome-wide distributions of TTF-1 binding regions and the transcriptional programs regulated by TTF-1 between NCI-H209 (H209), a human SCLC cell line, and NCI-H441 (H441), a human LADC cell line, using chromatin immunoprecipitation-sequencing (ChIP-seq) and RNA-sequencing (RNA-seq). TTF-1 binding regions in H209 and H441 cells differed by 75.0% and E-box motifs were highly enriched exclusively in the TTF-1 binding regions of H209 cells. Transcriptome profiling revealed that TTF-1 is involved in neuroendocrine differentiation in H209 cells. We report that TTF-1 and achaete-scute homolog 1 (ASCL1, also known as ASH1, an E-box binding basic helix-loop-helix transcription factor, and a lineage-survival oncogene of SCLC) are coexpressed and bound to adjacent sites on target genes expressed in SCLC, and cooperatively regulate transcription. Furthermore, TTF-1 regulated expression of the Bcl-2 gene family and showed antiapoptotic function in SCLC. Our findings suggest that TTF-1 promotes SCLC growth and contributes to neuroendocrine and antiapoptotic gene expression by partly coordinating with ASCL1.
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Insulin-transferrin-sodium selenite media supplement, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Trizma® base solution, 1.5 M