Merck
  • Identification of Moraxella bovis by using a monoclonal antibody to a lipopolysaccharide epitope.
Identification of Moraxella bovis by using a monoclonal antibody to a lipopolysaccharide epitope.
Journal of clinical microbiology (1989-12-01)
Y Wannemuehler, K Johansen, R Rosenbusch
ABSTRACT
A monoclonal antibody to the lipopolysaccharide of Moraxella bovis is described. In an indirect fluorescent-antibody test, this monoclonal antibody reacted with 39 of 39 strains of M. bovis tested and did not react with 26 nonfermenting gram-negative coccobacilli other than M. bovis. When used in an indirect fluorescent-antibody test, it proved useful for rapid and easy identification of M. bovis.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Pseudomonas aeruginosa 10, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Klebsiella pneumoniae, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides (rough strains) from Escherichia coli J5 (Rc mutant)