Merck
  • Synthesis, characterization and antitumour activity of metal complexes of 5-carboxy-2-thiouracil.
Synthesis, characterization and antitumour activity of metal complexes of 5-carboxy-2-thiouracil.
Metal-based drugs (2008-05-14)
U P Singh, S Singh, S M Singh
ABSTRACT
Metal complexes of 5-carboxy-2-thiouracil with Mn(ll), Co(ll), Ni(ll), Cu(ll), Zn(ll) and Cd(ll) ions were synthesized, characterized, and subjected to a screening system for evaluation of antitumour activity against Sarcoma-180 (S-180) tumour cells. The complexes were characterized by elemental analysis, infrared, electronic spectra, room temperature magnetic measurements and powder X-ray diffraction. The antitumour activity results indicate that some complexes have antitumour activity both in vivo and in vitro against S-180 tumour cells.
Sigma-Aldrich
5-Carboxy-2-thiouracil
Pricing and availability is not currently available.