Merck
  • Gene transfection efficiency of tracheal epithelial cells by DC-chol-DOPE/DNA complexes.
Gene transfection efficiency of tracheal epithelial cells by DC-chol-DOPE/DNA complexes.
Biochimica et biophysica acta (1999-07-17)
A Colosimo, A Serafino, F Sangiuolo, S Di Sario, E Bruscia, P Amicucci, G Novelli, B Dallapiccola, G Mossa
ABSTRACT
We evaluated the transfection efficiency of five different cationic liposome/plasmid DNA complexes, during the in vitro gene transfer into human epithelial tracheal cell lines. A dramatic correlation between the transfection efficiency and the charge ratio (positive charge of liposome to negative charge of DNA) has been found. DC-Chol-DOPE was found to be the most effective liposome formulation. Therefore, a morphological and structural analysis of DC-Chol-DOPE liposomes and DC-Chol-DOPE/DNA complexes, has been performed by transmission electron microscopy (TEM) and by confocal laser scanning microscopy (CLSM), respectively. The process of interaction between DC-Chol-DOPE/DNA complexes and human epithelial tracheal cells has been studied by CLSM. These results raise some issues for in vivo gene therapy.
Sigma-Aldrich
Cholesteryl 3β-N-(di­methyl­amino­ethyl)­carbamate hydrochloride, ≥95%
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder