Merck
  • Capsule-supporting ring: a new device for resin embedding of glass-mounted specimens.
Capsule-supporting ring: a new device for resin embedding of glass-mounted specimens.
Journal of microscopy (2009-04-29)
A Sawaguchi, F Aoyama, S Ide, T Suganuma
ABSTRACT
The goal of specimen preparation for transmission electron microscopy is to obtain high-quality ultra-thin sections with which we can correlate cellular structure to physiological function. In this study, we newly developed a capsule-supporting ring that can be useful for resin embedding of glass-mounted specimens. The present device allowed us to re-embed a semi-thin section on a microscope slide into a resin block not only for efficient ultra-thin sectioning but also for a correlative light and electron microscopy. Similar to epoxy resins for morphological observations, semi-thin sections of low-viscosity hydrophilic resins, such as Lowicryl series, can be re-embedded into the resin, which can be useful for cytochemical gold labelling. A further application of the present device improved flat embedding of cultured cells on glass cover slips for electron microscopy, preserving in situ sub-cellular structures close to their native state. We practically describe the use of capsule-supporting ring and demonstrate representative micrographs as results.
Sigma-Aldrich
Epoxy Embedding Medium kit, embedding resin for electron microscopy
Pricing and availability is not currently available.