
TWEEN® 80

Synonym(s):
POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80, TWEEN® 80
CAS Number:
Products
Product Category
Brand
Biological Source
Feature
Physical Form
Chemical Composition
Quality Segment
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing