
(Trimethylsilyl)methanesulfonate

Synonym(s):
Methanesulfonic acid trimethylsilyl ester, Trimethylsilyl mesylate
Linear Formula:
H3CSO3OSi(CH3)3
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing