
Tetrazole solution

Empirical Formula (Hill Notation):
CH2N4
CAS Number:
Molecular Weight:
70.05
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing