
Temozolomide Related Compound C

Synonym(s):
Diphenylcarbamoyl chloride, Chloroformic acid diphenylamide, Diphenylcarbamic chloride, NSC 6788
Linear Formula:
(C6H5)2NCOCl
CAS Number:
Molecular Weight:
231.68
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing