
Silicon Tetrabiphenyl MeOTAD

Empirical Formula (Hill Notation):
C104H88N4O8Si
CAS Number:
Molecular Weight:
1549.92
Products
Brand
Sustainability
Color
Feature
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing