Skip to Content
CF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Seleno-DL-cystine
Empirical Formula (Hill Notation):
C
6
H
12
N
2
O
4
Se
2
CAS Number:
2897-21-4
Molecular Weight:
334.09
Products
Product Category
amino acids (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Amino Acid
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
S1650
Seleno-DL-cystine,
>98% (TLC), suitable for HPLC
Expand
View Pricing