Skip to Content
Merck
(S)-(+)-Epichlorohydrin 98%

(S)-(+)-Epichlorohydrin

(S)-(+)-Epichlorohydrin 98%
Synonym(s):
(S)-(+)-2-(Chloromethyl)oxirane
Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing