Skip to Content
Merck
(S)-(+)-2-Methylpyrrolidine 95%

(S)-(+)-2-Methylpyrrolidine

(S)-(+)-2-Methylpyrrolidine 95%
Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing