
Rebaudioside F

Empirical Formula (Hill Notation):
C43H68O22
Molecular Weight:
936.99
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing