
Raspberry ketone

Synonym(s):
4-(4-Hydroxyphenyl)-2-butanone, Frambione, NSC 26515, Raspberry ketone
Linear Formula:
HOC6H4CH2CH2COCH3
CAS Number:
Molecular Weight:
164.20
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing