
Ranitidine Impurity J

Synonym(s):
1,1′-N-[Methylenebis(sulphanediylethylene)]bis(N′-methyl-2-nitroethene-1,1-diamine)
Empirical Formula (Hill Notation):
C11H22N6O4S2
CAS Number:
Molecular Weight:
366.46
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2