
Poly(N-isopropylacrylamide), carboxylic acid terminated

Synonym(s):
NIPAM polymer, PNIPAM-COOH, Polyacrylamide, functionalized polyNIPAM, functionalized polyacrylamide, polyNIPAM
Linear Formula:
(C6H11NO)nSCH2CH2CO2H
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Polymer Composition
Polymer Type