
Phenylephrine Related Compound G

Synonym(s):
(R)-N-[2-Hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylglycine, N-(Carboxymethyl)phenylephrine
Empirical Formula (Hill Notation):
C11H15NO4
CAS Number:
Molecular Weight:
225.24
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2