
Pentaethylenehexamine

Synonym(s):
3,6,9,12-Tetraazatetradecane-1,14-diamine, Pentaethylenehexamine
Linear Formula:
NH2CH2CH2NH(CH2CH2NH)3CH2CH2NH2
CAS Number:
Molecular Weight:
232.37
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing