
Oxydemeton-methyl Metabolite

Synonym(s):
O,O-Dimethyl S-[2-(ethylsulfinyl)ethyl]monothiophosphate, Demeton-S-methyl sulfoxide
Empirical Formula (Hill Notation):
C6H15O4PS2
CAS Number:
Molecular Weight:
246.28
Products
Brand
Formula Weight
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing