
Maslinic acid

Synonym(s):
2α,3β-Dihydroxyolean-12-en-28-oic acid, 2α-Hydroxyoleanolic acid, Crategolic acid
Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing