Skip to Content
Merck
JC-1 powder or solid (Crystals)

JC-1

JC-1 powder or solid (Crystals)
Synonym(s):
5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethyl-imidacarbocyanine iodide, 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethylbenzimidazolocarbocyanine iodide, CBIC2(3)
Empirical Formula (Hill Notation):
C25H27Cl4IN4
CAS Number:
Molecular Weight:
652.23

Products

Product Category
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing