
Iohexol Related Compound A

Synonym(s):
5-(Acetylamino)-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide, 5-Acetylamino-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide
Empirical Formula (Hill Notation):
C16H20I3N3O7
CAS Number:
Molecular Weight:
747.06
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing