
Indoxacarb

Synonym(s):
Methyl 7-chloro-2,5-dihydro-2-[N-(methoxycarbonyl)-4-(trifluoromethoxy)anilinocarbonyl]indeno[1,2-e][1,3,4]oxadiazine-4a(3H)carboxylate
Empirical Formula (Hill Notation):
C22H17ClF3N3O7
CAS Number:
Molecular Weight:
527.83
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2