![Hypotaurine ≥98% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/159/041/3af61dd5-a2ed-40b6-9c8f-e1177ff2f84c/640/3af61dd5-a2ed-40b6-9c8f-e1177ff2f84c.png)
Hypotaurine
![Hypotaurine ≥98% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/159/041/3af61dd5-a2ed-40b6-9c8f-e1177ff2f84c/640/3af61dd5-a2ed-40b6-9c8f-e1177ff2f84c.png)
Synonym(s):
2-Aminoethanesulfinic acid
Empirical Formula (Hill Notation):
C2H7NO2S
CAS Number:
Molecular Weight:
109.15
Products
Product Category
Brand
Color
Formula Weight
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing