
Hyoscine hydrobromide impurity B

Synonym(s):
(1R,2R,4S,5S,7s)-3-Oxa-9-azatricyclo[3.3.1.02,4]non-7-yl (2S)-3-hydroxy-2-phenylpropanoate, Norhyoscine, Norscopolamine
Empirical Formula (Hill Notation):
C16H19NO4
CAS Number:
Molecular Weight:
289.33
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing