Skip to Content
CF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
GlycylGlycine
Synonym(s):
Gly-Gly, Diglycine, Glycyl-glycine
Linear Formula:
NH
2
CH
2
CONHCH
2
COOH
CAS Number:
556-50-3
Molecular Weight:
132.12
EC Number:
209-127-8
Products
Product Category
buffers (1)
Brand
SAFC (1)
Biological Source
Formula Weight
pH Value
Physical Form
pK Value
Quality Segment
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
G0674
GlycylGlycine,
Expand
View Pricing