
Glycopyrrolate Related Compound L

Synonym(s):
Methyl 2-cyclopentyl-2-hydroxy-2-phenylacetate, Cyclopentylmandelic acid methyl ester, Methyl cyclopentylphenylglycolate, NSC 93811
Empirical Formula (Hill Notation):
C14H18O3
CAS Number:
Molecular Weight:
234.29
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2