
Glass sand

Synonym(s):
Sand, white quartz, Quartz, Silicon dioxide
Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
Products
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing