
Formoterol fumarate

Synonym(s):
Formoterol fumarate dihydrate, ((±)-2′-Hydroxy-5′-[(R*)-1-hydroxy-2-[[(R*)-p-methoxy-a-methylphenethyl]amino]ethyl]formanilide fumarate (2:1) (salt) dihydrate
Empirical Formula (Hill Notation):
(C19H24N2O4)2 · C4H4O4 · 2H2O
CAS Number:
Molecular Weight:
840.91
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade