
Fluvoxamine for system suitability

Synonym(s):
Fluvoxamine maleate, (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone-O-(2-aminoethyl)oxime maleate
Empirical Formula (Hill Notation):
C15H21F3N2O2 · C4H4O4
CAS Number:
Molecular Weight:
434.41
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing