
Fluticasone propionate Related Compound J

Synonym(s):
6α, 9α-Difluoro-11β,17α-dihydroxy-16α-methyl-3-oxoandrosta-1,4-diene-17β-carboxylic acid
Empirical Formula (Hill Notation):
C21H26F2O5
CAS Number:
Molecular Weight:
396.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing