
Fluticasone propionate for impurity C identification

Synonym(s):
Fluticasone propionate, (6α,11β,16α,17α)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)androsta-1,4-diene-17-carbothioic acid S-(fluoromethyl) ester
Empirical Formula (Hill Notation):
C25H31F3O5S
CAS Number:
Molecular Weight:
500.57
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing