
Fluorescent Brightener 28 disodium salt solution

Synonym(s):
Fluostain I, Calcofluor white M2R, Tinopal LPW
Linear Formula:
C40H42N12O10S2Na2
CAS Number:
Molecular Weight:
960.95
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing