
Ensulizole

Synonym(s):
2-Phenyl-5-benzimidazolesulfonic acid, Ensulizole
Empirical Formula (Hill Notation):
C13H10N2O3S
CAS Number:
Molecular Weight:
274.30
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing