
Enilconazole

Synonym(s):
Imazalil, 1-[2-(Allyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole
Empirical Formula (Hill Notation):
C14H14Cl2N2O
CAS Number:
Molecular Weight:
297.18
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing