
DMT-locA(bz) Phosphoramidite

Synonym(s):
DMT-locA(bz) Phosphoramidite
Empirical Formula (Hill Notation):
C48H52N7O8P
CAS Number:
Molecular Weight:
885.94
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-3 of 3
Compare
Product Number
Description
Pricing