(±)-Norpropoxyphene-d5 maleate solution
Empirical Formula (Hill Notation):
C21D5H22NO2 · C4H4O4
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing