
D-α-Hydroxyglutaric acid-13C5 disodium salt

Synonym(s):
2-Hydroxyglutarate-13C5, (R)-2-Hydroxypentanedioic acid-13C5 disodium salt, Disodium (R)-2-hydroxyglutarate-13C5
Empirical Formula (Hill Notation):
13C5H6Na2O5
CAS Number:
Molecular Weight:
197.04
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing