Skip to Content
Merck
Cyclic adenosine diphosphate-ribose ≥90% (HPLC), lyophilized powder

Cyclic adenosine diphosphate-ribose

Cyclic adenosine diphosphate-ribose ≥90% (HPLC), lyophilized powder
Synonym(s):
ADP-cyclo[N1:1′′]-ribose, Cyclic ADPR, cADP-ribose, cADPR
Empirical Formula (Hill Notation):
C15H21N5O13P2
CAS Number:
Molecular Weight:
541.30

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing