
Cyanidin 3-arabinoside chloride

Synonym(s):
Cyanidin 3-O-α-L-arabinoside chloride, Cyanidin 3-α-L-arabinopyranoside, Cyanidin 3-α-L-arabinoside, Cyanidin 3-arabinoside chloride
Empirical Formula (Hill Notation):
C20H19ClO10
CAS Number:
Molecular Weight:
454.81
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing